Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
P0834
Sigma-Aldrich
Protein standard
liquid, 2 mg protein/ml
Synonym
Albumin from bovine serum, protein standard
CAS Number
9048-46-8
MDL number
MFCD00130384
Expand/Collapse All
Price and Availability
Descriptions
Application
Concentrated solution for all protein assays.
Packaging
Sealed ampules provide assurance that the concentration will be correct with every assay.
Physical form
2 mg bovine serum albumin/ml in 0.9% NaCl containing 0.05% sodium azide
Properties
form
liquid
color
clear, colorless
storage temp.
2-8°C
Safety
WGK Germany
3
Related Categories
...
Plant Proteomics
>
Protein Quantitation
...
General Protein Quantitation
>
Protein Standards
Scroll Up
Scroll Down
Related Information
Plant Biotech Guide (163kb)
MSDS
Specification Sheet
Certificate of Analysis
Certificate of Origin
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
P0834 (Sigma-Aldrich)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.