Application
A differentiated cell line derived from H4-11-E-C3 (ECACC catalog number 85061112). A versatile tool for studying regulation of mono-oxygenase activities and analyzing their role in the activation of xenobiotics, e.g. carcinogens, drugs, pesticides.
Subculture: Spit at 70-80% confluency, approx. 1:3 to 1:6 (2-4 × 104 cells/cm2) Trypsinize using 0.25% solution, with or without EDTA, 37 oC and 5% CO2.
Recommended culture medium: Coon's Modified Ham's F12 + 2 mM L-Glutamine + 10% FBS or Kaign's Modified Ham's F12 + 2 mM L-Glutamine + 45 mg/L ascorbic acid + 18 mg/L myo-inositol + 10% FBS.