Components
contains:
Fluorescence Spectral Standard A
Fluorescence Spectral Standard B
Fluorescence Spectral Standard C
Fluorescence Spectral Standard D
Fluorescence Spectral Standard E
Ethanol Spectral
BAM-Certificate
CD
Application
suitable for the spectral range of 300-760 nm
General description
With the Kit Spectral Fluorescence Standards, we provide a simple, flexible, and traceable calibration tool for the determination and control of the relative spectral responsivity of fluorescence instruments. The Kit is optimized for spectrofluorometers and can be employed for a broad variety of measurement geometries. With proper consideration of the underlying measurement principle, it can be adapted to the calibration of other types of fluorescence instruments.