![]() |
Synonym
Monoclonal Anti
Descriptions
Preparation Note
Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).
Specificity
The product recognizes epitopes located on the rat κ and λ light chains on the various rat immunoglobulin classes and subclasses. The epitope recognized on the κ light chains is shared by both the 1a and 1b allotypes of this molecule. The antibody detects the κ and λ light chains derived from normal serum immunoglobulins or from myeloma proteins. Weak cross-reaction is observed with guinea pig immunoglobulins but not with human, bovine, cat, chicken, dog, goat, horse, mouse, pig, rabbit or sheep immunoglobulins.
Physical form
Lyophilized from a solution containing 1% BSA and 0.01% thimerosal
General description
The product is a mixture derived from hybridomas RT-39 and RL-6, secreting monoclonal antibodies against rat κ and λ light chains, respectively.
Properties
antibody form
purified immunoglobulin
clone
RT-39/RL-6
form
lyophilized powder
mol wt
antigen mol wt 25 kDa
packaging
vial of 0.5 mL
application(s)
direct ELISA: 1:40,000
dot blot: 1:4,000
immunoblotting (chemiluminescent): 1:20,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
isotype
IgG1/IgG2a
storage temp.
2-8°C
| ||||||||