![]() |
Synonym
Anti
Anti
Anti
Anti
Anti
Anti
Anti
Monoclonal Anti
Properties
antibody form
purified immunoglobulin
clone
35C1
form
buffered aqueous solution
mol wt
antigen mol wt ~46 kDa
species reactivity
human, mouse
concentration
~2 mg/mL
application(s)
immunoblotting: suitable
immunocytochemistry: 2.5-5 μg/mL using starved HeLa cells treated with 10% fetal calf serum
indirect ELISA: suitable
isotype
IgG2b
Gene Information
mouse ... Aurka(20878)
human ... AURKA(6790)
shipped in
dry ice
storage temp.
−20°C
References
Reference
Krystyniak, A., et al., Inhibition of Aurora A in response to DNA damage. Oncogene 25, 338-348, (2006)
Cremet, J.Y., et al. Mol. Cell Biochem. 243, 123-131, (2003) abstract
Cremet, J.Y., et al., Preparation and characterization of a human Aurora-A kinase monoclonal antibody. Mol. Cell Biochem. 243, 123-131, (2003) abstract
Krystyniak, A., et al., Inhibition of Aurora A in response to DNA damage. Oncogene 25, 338-348, (2006)
Dutertre, S., et al., Phosphorylation of CDC25B by Aurora-A at the centrosome contributes to the G2-M transition. J. Cell Sci. 117, 2523-2531, (2004) abstract
Dutertre, S., et al. J. Cell Sci. 117, 2523-2531, (2004) abstract
| ||||||||