Compatibility
Compatibility
Corning 4961 2-200 μl
Eppendorf 4810 2-20 μl
Eppendorf Research/Reference 2-20 μl
Eppendorf Research Pro 4860 5-100 μl
Eppendorf Titerman 4908 5-50 μl
Hamilton SoftGrip Precision 25 μl
Nichriyo/Oxford BenchMate 2-20 μl
Pipetman P-20 2-20 μl
Rainin EDP 2 2.5-25 μl
Rainin EDP Plus 2.5-25 μl
Rainin Pipet Plus 2.5-20 μl
Rainin Pipetman 2.5-20 μl
Socorex/Wheaton Calibra/822 2-20 μl
Transferpette 2-20 μl
Transferpette Multi-Channel (Limited Draw) 2.5-25 μl/5-50 μl
General description
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
10 μL reference mark; well suited for low volumes.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.