Compatibility
Compatibility
Biohit mLINE 100-1000 μl
Capp Aero 1000 μl
Corning 4964 100-1000 μl
Eppendorf Research Pro 50-1000 μl
Eppendorf Response 4850 50-1000 μl
Finnpipette 4027/4500 Digit 200-1000 μl
Nichiryo BenchMate 100-1000 μl
Pipetman P-1000 100-1000 μl
Rainin EDP 2 100-1000 μl
Rainin EDP Plus 100-1000 μl
Rainin Pipet Plus 100-1000 μl
Rainin Pipetman 100-1000 μl
Socorex/Wheaton Calibra/822 100-1000 μl
Transferpette 100-1000 μl
General description
Precision-molded tips with the patented Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.