General description
Handheld, single-use Osborn Scientific Biowarfare Agent Detection Devices™ for qualitative environmental testing for Anthrax. Accurately detects minute amounts of Anthrax on surfaces or in solutions in as little as 3 minutes. Tests are individually packaged in a foil pouch along with the simulant, pipette and swab. No additional accessories needed to perform an immediate evaluation, anywhere in the world.
• Detection Level - As little as 25 micrograms of Anthrax
• Sensitivity - 100% accurate as independently tested by the Department of Defense
• Specificity - Testing of substances evaluated produced no false positives or negatives
• Easy to use - Completely self contained
Legal Information
Biowarfare Agent Detection Device is a trademark of Osborne Scientific Group.