antibody form
affinity isolated antibody
form
lyophilized powder
mol wt
antigen mol wt 110 kDa
species reactivity
human, mouse, rat, chicken, frog, bovine, pig, fish, nonhuman primates, hyena, hamster, vole, ferret
application(s)
immunoblotting: 0.5-2 μg/mL using RIPA lysates from human LNCap lysate
immunocytochemistry: 10 μg/mL using LNCp cells fixed with 4% paraformaldehyde and permeabilized with 0.1% Triton X-100
immunoprecipitation: suitable
Gene Information
shipped in
wet ice
storage temp.
2-8°C