![]() |
Synonym
Anti
Anti
Anti
Anti
Anti
Properties
antibody form
purified immunoglobulin
clone
A3C1
form
buffered aqueous solution
mol wt
antigen mol wt ~220 kDa
species reactivity
human
concentration
~2 mg/mL
application(s)
immunoblotting: 0.5-1 μg/mL using Helatotal cell extract
immunocytochemistry: suitable
immunoprecipitation: suitable
microarray: suitable
isotype
IgG1
Gene Information
human ... NCOA6(23054)
shipped in
dry ice
storage temp.
−20°C
| ||||||||