General description
The new AlphaDigiDoc RT 2 digital imaging system provides easy to use real time sample positioning with a compact space requirement set at an affordable cost that fits most laboratory budgets. AlphaDigiDoc RT 2 is paired with the powerful AlphaEaseFC Imaging Analysis software that combines unrivaled ease of use with a comprehensive set of documentation, analysis and image enhancement tools. The real time, high resolution gel documentation system is the result of feedback from thousands of Alpha Innotech users!
• Real time gel documentation, fluorescence and absorbance imaging!
• Easy connection to any computer's USB port
• Flexible design with multiple light source and darkroom cabinet options
Key features of AlphaDigiDoc RT 2 System include:
• Real Time Imaging for ease of use and fast sample positioning
• Create vibrant images with up to 8-megapixel resolution in 8-bit grayscale or 24-bit color images of exceptional quality
• 4x optical zoom and autofocus via software control
• A compact darkroom hood makes AlphaDigiDoc RT 2 System ideal for labs with limited bench space, while the advanced MultiImage II light cabinet provides unmatched flexibility and upgradeability.
• Use your existing desktop orlaptop computer and UV transilluminator without installing ANY additional hardware.
• AlphaEase FC image acquisition software delivers zoom lens, resolution and exposure control with "intelligent" MultiImage® II cabinet controls.
• AlphaEase FC automatic image enhancement and data managment tools provide a comprehensive set of features for a variety of scientific applications.
• Large format thermal and color dye-sublimation digital printer option generates hard copies in seconds.