![]() |
Properties
antibody form
affinity isolated antibody
form
buffered aqueous solution
species reactivity
human
application(s)
immunoblotting: 1:2,000 using human AFX expressed in transfected COS-7 cell extracts
indirect immunofluorescence: 1:1,000 using paraformaldehyde-fixed AFX transfected COS-7 cells
microarray: suitable
Gene Information
human ... FOXO4(4303)
shipped in
dry ice
storage temp.
−20°C
| ||||||||