Application
Prior to reaction with alkaline phosphatase, the BCIP reagent is a colorless to faint blue solution, and the INTX reagent is a colorless to light yellow solution. The two component mixture develops a orangish-red product when reacted with alkaline phosphatase in membrane type assays. This substrate is not recommended for ELISA assays.
Physical form
Sufficient for preparing 400 mL of working substrate.
Supplied as a 2 component buffered alkaline phosphatase substrate containing BCIP and INTX