General description
All the power of a laboratory balance in a portable unit. Setup is simple with 4 cursor keys, straightforward menus, and prompts that easily guide you through the process. Designed with contours for easy cleaning, sealed front panel and a spill gutter to direct liquids off the balance. Easy calibration, AC adapter supplied or standard alkaline batteries (8 AA required).
Standard Features:
• RS232 bidirectional interface
• auto shut off, weigh below, integral security bracket
• rugged composite housing with sealed front panel and stainless steel pans
• tactile feedback switches
• user selectable stability levels and filters
• stability indicator
• menu and calibration lock-out switch.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.