![]() |
Properties
antibody form
affinity isolated antibody
form
buffered aqueous solution
mol wt
antigen mol wt 200 kDa
species reactivity
human
application(s)
immunoblotting: 1:4,000-1:8,000 using nuclear extracts of HeLa cells.
indirect immunofluorescence: 10-20 μg/mL using CHO cells fixed with paraformaldehyde/Triton.
microarray: suitable
Gene Information
mouse ... Smarca4(20586)
human ... SMARCA4(6597)
shipped in
dry ice
storage temp.
−20°C
| ||||||||