antibody form
affinity isolated antibody
form
lyophilized powder
mol wt
antigen mol wt 16-18 kDa (a double band using recombinant mouse Blk)
antigen mol wt 27 kDa (from mouse tissue extracts of seminal vesicles, kidney, and liver)
species reactivity
mouse
application(s)
immunoblotting (chemiluminescent): 2 μg/mL using extracts from E. coli cells expressing mouse Blk (Bcl2-interacting Killer-like) and extracts from mouse seminal vesicles, kidney, and liver.
microarray: suitable
Gene Information
storage temp.
−20°C