General description
• Amplification Labtop Cooler protects enzymes and solutions by maintaining freezer temperature
• Temperature is maintained between -20 °C and - 15 °C during sample preparation
• Protects valuable reagents in the freezer from fluctuations due to defrost cycles, open doors or power failure.
• The aluminum block holds up to 96 each 0.2 mL tubes in an 8 × 12 array.
• Compatible with GeNunc 0.2 mL individual tubes and strips, 96 well Amplification Plate and accommodates GeNunc Tube Tray