General description
Peel-and-stick adhesive enclosures isolate specimens on glass microscope slides. Chamber design minimizes friction, promotes reagent mixing and facilitates uniform hybridization. Sealable ports in the chamber surface allow for addition and removal of reactants. Leak-proof chambers are temperature resistant and eliminate evaporation. Secure-Seal adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating. RNase-free and ready to use.
Legal Information
SecureSeal is a trademark of Grace Bio-Labs, Inc.