General description
Like Colorburst™ and Chemichrome™ Western Control, Chemichrome Ultimate contains eight brilliantly colored polypeptides which serve as positive controls during protein migration and transfer. In addition to mouse IgG, Chemichrome Ultimate also contains goat and rabbit IgG, which serve as positive controls for Western blotting using either colorimetric or chemiluminescent substrates.
Legal Information
Chemichrome is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.