General description
Researchers review protein chemistry and behavior of this class of proteins, and provide experimental techniques for studying it both in vitro and in vivo. This second volume focuses on techniques for studying the solution structure, stability, dynamics, calcium-binding properties, and biological activity of calcium-binding proteins in general. In addition to enzymatic assays and more routine spectroscopic and protein chemistry techniques, there are also NMR approaches, thermodynamic analyses, kinetic measurements such as surface plasmon resonance, strategies for amino acid sequence alignments, and fluorescence methods to study the distribution of calcium and calcium-binding proteins in cells.
Table of Contents
Methods to Study Calcium-Binding Proteins Quantitative Analysis of Ca2+ Binding by Flow Dialysis
Calcium Binding to Proteins Studied via Competition with Chromophoric Chelators Deconvolution of Calcium Binding Curves: Facts and Fiction Absorption and Circular Dichroism Spectroscopy S Fourier Transform Infrared Spectroscopy of Calcium-Binding Proteins Steady State Fluorescence Spectroscopy Fluorescence Methods for Measuring Calcium Affinity and Calcium Exchange with Proteins Surface Plasmon Resonance of Calcium-Binding Proteins Differential Scanning Calorimetry Isothermal Titration Calorimetry Multiangle Laser Light Scattering and Sedimentation Equilibrium Small-Angle Solution Scattering Reveals Information on Conformational Dynamics in Calcium-Binding Proteins and in Their Interactions with Regulatory Targets Investigation of Calcium-Binding Proteins Using Electrospray Ionization Mass Spectrometry Synthetic Calcium-Binding Peptides Gary S. Shaw Proteolytic Fragments of Calcium-Binding Proteins Electron Magnetic Resonance Studies of Calcium-Binding Proteins Cadmium-113 and Lead-207 NMR of Calcium-Binding Proteins Calcium-43 of NMR of Calcium-Binding Proteins Exploring Familial Relationships Using Multiple Sequence Alignment Structure Determination by NMR - Isotope Labeling Protein Structure Calculation from NMR Data Shape and Dynamics of Calcium Binding Proteins Investigated by Nitrogen-15 NMR Relaxation The Use of Dipolar Couplings for the NMR Structure Refinement of a Pair of Calcium Binding EGF Domains Vector Geometry Mapping: A Method to Characterise the Conformation of Helix-Loop-Helix Calcium Binding Proteins Use of Calmodulin Antagonists and S-100 Protein Interacting Drugs for Affinity Chromatography Enzymatic Assays to Compare Calmodulin Isoforms, Mutants and Chimeras Gene Expression in Transfected Cells Monitoring the Intracellular Free Ca2+-Calmodulin Concentration with Genetically-Encoded Fluorescent Indicator Proteins Studying the Spatial Distribution of Ca2+-Binding Proteins: How Does it Work for Calmodulin?
| ||||||||