Analysis Note
Measured by its ability to induce cytochrome c release from isolated mouse liver mitochondria.
Biochem/physiol Actions
Caspase-8-cleaved BID relocates from the cytosol to the outer mitochondrial membrane where its interaction with Bak alters mitochondrial membrane permeability.
Physical form
0.2 μm filtered solution in 25 mM HEPES, pH 7.5, and 0.1 M KCl.
General description
Human BID is cleaved with caspase-8 to generate an amino-terminal fragment (amino acids 1-60, 7 kDa) and the C-terminal fragment (amino acids 61-195, 15 kDa). On size exclusion chromatography cleaved BID elutes as a single band corresponding to 27 kDa indicating that the two fragments remain associated.