General description
NovaChamber is a tube rack with cooling capability. Water can be frozen inside the rack, chilling the sturdy polypropylene collar which houses the sample tubes. A snap-on bottom cover is designed to seal the liquid inside. NovaChamber accommodates all commonly used tube sizes: 24 microcentrifuge tubes (0.5-2 mL), 14 tubes (12x75 mm culture tube, cryovial and a variety of blood tubes), 7 tubes (15 mL centrifuge tube) and one 50 mL tube. The cooling can be regenerated by simply placing the NovaChamber in the freezer to refreeze the liquid inside.
Lower temperature limit is 0șC. Not for use below 0 C.
Legal Information
Novachamber is a trademark of Diversified Biotech.