General description
Chambered coverglass products consist of removable / reusable non-cytotoxic silicone gaskets secured to 24 mm x 50 mm, number 1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.
• The thin flat coverglass is the optimal platform for high resolution microscopy an high-sensitivity, quantitative, fluorescent imaging.
• Leak-proof design eliminates well-to-well cross contamination.
• Gaskets stabilize coverglass and provide convenient handling using conventional microscope slide holders and accessories.
• Gaskets are removable for high resolution imaging and may be re-sterilized and reused.
• Well spacing facilitates liquid handling utilizing multichannel pipettes and robotic dispensers.