grade
SigmaUltra
assay
≥99.0% anhydrous basis (CAPSO, titration)
≥99.0% anhydrous basis (TSOD, titration)
total impurities
≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter
≤1.0% (Karl Fischer)
ign. residue
≤0.1%
pH
2.0-4.0 (20 °C, 0.5 M in H2O)
useful pH range
8.9 - 10.3
pKa (25 °C)
9.6
solubility
H2O: 0.5 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%
cation traces
Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%
abs.
A0.5M/260, H2O ≤0.05
A0.5M/280, H2O ≤0.05