![]() |
Descriptions
Immunogen
synthetic peptide corresponding to amino acids 423-440 of human CtBP1. The corresponding sequences in mouse and rat differ by one and two amino acids, respectively. The peptide does not cross-react with CtBP2.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Properties
antibody form
affinity isolated antibody
form
buffered aqueous solution
mol wt
antigen mol wt 48 kDa
species reactivity
human, mouse
concentration
~1 mg/mL
application(s)
immunoblotting: 0.5-1 μg/mL using extracts of K-562 cells
immunoblotting: 1-2 μg/mL using mouse brain extracts
immunoprecipitation: 5-10 μg using extracts of K-562 cells.
microarray: suitable
Gene Information
human ... CTBP1(1487)
mouse ... Ctbp1(13016)
rat ... Ctbp1(29382)
shipped in
dry ice
storage temp.
−20°C
| ||||||||