Application
Solutions of Cycloheximide and Actinomycin D to be used as enhancers of death receptor induced apoptosis and a solution of Staurosporine, a wide spectrum apoptosis inducer, as a positive control are included. Apoptosis can be determined by measuring caspase 3 activity, using the Sigma CASP-3F kit in a high throughput format.
Biochem/physiol Actions
Apoptotic processes involve well-orchestrated interactions of cell death receptor (CD95-Fas, TNF-R), death receptor adaptors (FADD, TRADD), caspases, Bcl-2 family members and other proteins. Some cells, such as HeLa, HepG2, NT68, rat mesangial cells, and thyroid follicular cells, are resistant to death receptor mediated apoptosis. Maintaining the state of the resistance often requires de novo protein or RNA synthesis. Therefore, treatment with protein synthesis inhibitors, such as cycloheximide (CHX), or RNA synthesis inhibitor, such as actinomycin D (ActD) can sensitize resistant cells. CHX and ActD can also be used as a tool to investigate the apoptosis inhibitors in these cells.