General description
Corning RoboFlask vessels are robotics compatible cell culture flasks offering 92.6 cm2 cell growth surface area. The flasks are designed for use in automated cell culture systems utilizing a microplate-size format.
• Removable septum cap for both automated systems and manual applications
• Septum cap ensures sterility through multiple piercings
• Microplate-size footprint
• Bar coded
• Compatible with the Tecan Cellerity Cell Maintenance and Assay System
• Built-in vent membrane for efficient gas and liquid exchange
Legal Information
Corning is a registered trademark of Corning, Inc.
RoboFlask is a trademark of Corning, Inc.