General description
Corning offers a complete polypropylene storage system consisting of plates, blocks, and mats. All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 384 well polypropylene plates and blocks are built to proposed industry standard footprint. All plates and blocks are automation-friendly and solvent resistant. The 384 well plates and blocks are certified DNase/RNase free and feature rounded bottoms for optimal liquid handling and a rounded square-well geometry for reduced wicking.
Legal Information
Corning is a registered trademark of Corning, Inc.