General description
The Thermowell PCR plates are designed for use in most thermal cyclers and offer a rigidity that makes them ideal for automation. Each plate is 100% integrity-tested to ensure quality and reliability, and each lot is tested and certified to be RNase and DNase free. They also offer an extremely uniform wall thickness which enables heat to transfer quickly and consistently from well to well
Legal Information
Corning is a registered trademark of Corning, Inc.
THERMOWELL is a registered trademark of Corning, Inc.