General description
These low profile flasks have 33% greater growth area, in 33% less height than a standard T75 flask. 3 of these flasks take up only half the incubator space of 4 T75 flasks, effectively doubling incubator growth capacity. Their slim profile and 1/2 turn cap makes them easy to handle, and the angled neck design gives direct access to corners with a 10ml pipet
• 100 cm² available growth area
• Manufactured from optically clear virgin polystyrene.
• 100% integrity tested.
• Sterilized by γ-irradiation and certified nonpyrogenic
Legal Information
Corning is a registered trademark of Corning, Inc.