General description
Corning epoxide coated slides provide the optimal, uniform surface chemistry for covalent attachment of unmodified or amino-modified short oligonucleotides (~ 30-mer), as well as long oligonucleotides (>50-mer) and cDNA. The Pronto! Epoxide Spotting Solution when used in conjunction with Corning Epoxide Coated Slides, provides superior spot size control of printing high density arrays.
• Interslide CVs below 10 %
• Detect 1 pg RNA spiked into 4µg of total RNA sample
• Differentiate between 90% homologous oligos (3 mismatches in 30-mer oligonucleotides)
Legal Information
Corning is a registered trademark of Corning, Inc.