General description
The amino silane surface on UltraGAPS slides is ideal for printing long (>50mer) oligonucleotides. The added step to the manufacturing process means the UltraGAPS slide has a more hydrophobic surface GAPS II, resulting in smaller spots. Quality Certificate is included in each box of slides. The results of nine quality parameter are listed including DNA retention and an image of the spot morphology for that lot of slides.
Legal Information
UltraGAPS is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.