Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
D2690
Sigma
Dansyl alginate
>95%, powder
Synonym
Alginic acid, dansylated
MDL number
MFCD00676817
Expand/Collapse All
Price and Availability
Properties
assay
>95%
form
powder
mol wt
mol wt ~350,000 Da
color
yellow
solubility
H
2
O: soluble
storage temp.
2-8°C
Safety
WGK Germany
3
Related Categories
...
Amino Acid Analysis
>
Amino Acid Analysis Reagents
...
Fluorescent Conjugates
>
Amino acids and other organic acid conjugates
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
D2690 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.