Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
D7657
Sigma
Anti-Digitoxin antibody produced in rabbit
whole antiserum
MDL number
MFCD00163469
Expand/Collapse All
Price and Availability
Descriptions
Immunogen
Digitoxin-BSA.
Reconstitution
Dilute with buffer: 0.01 M phosphate, pH 7.4, 0.15 M sodium chloride and 0.1% sodium azide.
Properties
antibody form
whole antiserum
contains
15 mM sodium azide
application(s)
radioimmunoassay: 1:4,000
shipped in
dry ice
storage temp.
−20°C
Safety
WGK Germany
3
Related Categories
...
Antibodies for Neurobiology
>
Antibodies to Pharmacologicals
...
Alphabetical Index
>
D
Scroll Up
Scroll Down
Related Information
D7657 - Datasheet (27kb)
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
D7657 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.