Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
DMB10
Sigma
Percoll
®
density marker bead kit
Expand/Collapse All
Price and Availability
Descriptions
Application
For calibration of density gradients of Percoll
Legal Information
Percoll is a registered trademark of GE Healthcare.
Related Categories
...
Hematology and Histology
>
Cell Separation (Centrifugation) Media
...
Cell Fractionation and Organelle Isolation
>
Centrifugation
Scroll Up
Scroll Down
Related Information
DMB10 - Bulletin (24kb)
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
DMB10 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.