Analysis Note
The biological activity is measured by its ability to induce chemotaxis of rat Y3 cells transfected with the CCR3 receptor.
Physical form
Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA containing 1.25 mg bovine serum albumin.
Biochem/physiol Actions
Eotaxin-2 is a potent eosinophil chemotactic factor, which is involved in homeostatic, allergen-induced, and IL-4 related immune responses. Eotaxin-2 exhibits 40%, 42%, and 39% amino acid identity to MCP-3, MIP-1α, and eotaxin, respectively. Mature mouse and human eotaxin-2 share approximately 57% amino acid sequence identity.