General description
A UV-transmittable sheet can provide reliable protection against scratches and nicks to the surface of a transilluminator's filter glass. Fluoro-Sheet provides a two-dimensional fluorescent scale. This improved versatility provides researchers a better way to assess the distance of DNA migrated in the gel, since the rate of DNA migration is not always the same on each side of the gel. It is designed for UV transilluminators of medium (300 nm) and long (365 nm) wavelengths. Does NOT transmit UV of short wavelength (260 nm).
Other Notes
For 20 × 19 cm gels