General description
Kodak Professional Films provide the highest quality for accurate scientific imaging. While consumer films provide clear, sharp photographs, they are manufactured with a color-balance bias to allow for emulsion maturation in storage. Professional demands for highly accuate color rendition cannot tolerate this bias. Professional film, therefore, is at or near its color-balance optimum when manufactured. The film will stay near this balance when it is stored at or below 55 °F (13 °C) and processed promptly after exposure - before the expiration date printed on the carton.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company.