![]() |
Synonym
Monoclonal Anti
Descriptions
Specificity
Recognizes an epitope located on the γ (heavy)-chain of rabbit IgG. In immunoblotting, the antibody recognizes both native and denatured forms of rabbit IgG. In ELISA, the antibody is specific for rabbit IgG, and shows no cross-reactivity with rabbit IgA and IgM or human IgG, IgA, and IgM. No cross reaction is observed with IgG from the following species: bovine, cat, chicken, dog, goat, guinea pig, horse, pig, rat, or sheep.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.
Properties
antibody form
purified immunoglobulin
clone
RG-96
form
buffered aqueous solution
does not react with
human, guinea pig, rat, bovine, goat, sheep, horse, canine, feline, pig, chicken
application(s)
dot immunobinding: 1:64
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20
particle immunofluorescence: 1:32
isotype
IgG1
shipped in
dry ice
storage temp.
−20°C
| ||||||||