General description
Medium contrast; coaterless; high resolution print.
These instant films are Print Only (P) or Print-plus-Negative (P/N). The negatives require brief clearing in sodium sulfite solutions. Unless otherwise noted, prints need to be coated. Films have panchromatic spectral sensitivity. Professional equivalents have the same characteristics as their counterparts, but meet more stringent photographic specifications.
Legal Information
Polaroid is a registered trademark of Polaroid Corp.