Application
Clarification of serum
Ultra-low protein binding (<1 μg/cm2). Component materials meet USP XXI Class VI Biological Tests for plastics. Water extractables <0.5%
General description
Units contain Durapore Hydrophilic PVDF filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings. Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 ml; the 13 mm units are recommended for volumes of 0.5 - 10 ml.
Legal Information
Durapore is a registered trademark of Millipore Corp.
Millex is a registered trademark of Millipore Corp.