Construction of Microsatellite-Based, High-Resolution Genetic Maps in the Mouse, .
Genetic Analysis of Complex Traits, .
Sequence-Based Detection of Single Nucleotide Polymorphisms,
Genomic Mismatch Scanning for the Mapping of Genetic Traits,
Detection of Chromosomal Abnormalities by Comparative Genomic Hybridization,
Construction of a Bacterial Artificial Chromosome Library,
Contiguation of Bacterial Clones, .
Mapping of Genomic Clones by Fluorescence In Situ Hybridization,
Map Integration: From a Genetic Map to a Physical Gene Map and Ultimately to the Sequence Map,
Construction of Full-Length–Enriched cDNA Libraries: The Oligo-Capping Method,
Construction of Transcript Maps by Somatic Cell/Radiation Hybrid Mapping: The Human Gene Map, .
Preparation and Screening of High-Density cDNA Arrays with Genomic Clones, .
Direct Selection of cDNAs by Genomic Clones,
Exon Trapping: Application of a Large-Insert Multiple-Exon-Trapping System,
Sequencing Bacterial Artificial Chromosomes, .
Finding Genes in Genomic Nucleotide Sequences by Using Bioinformatics,
Gene Identification Using the Pufferfish, Fugu rubripes, by Sequence Scanning,
Isolation of Differentially Expressed Genes Through Subtractive Suppression Hybridization,
Isolation of Differentially Expressed Genes by Representational Difference Analysis, .
Expression Profiling and the Isolation of Differentially Expressed Genes by Indexing-Based Differential Display,
Expression Profiling by Systematic High-Throughput In Situ Hybridization to Whole-Mount Embryos,
Expression Monitoring Using cDNA Microarrays: A General Protocol, ,
Prediction of Protein Structure and Function by Using Bioinformatics,
Identification of Novel Genes by Gene Trap Mutagenesis,
Determination of Gene Function by Homologous Recombination Using Embryonic Stem Cells and Knockout Mice,
Genomic Analysis Utilizing the Yeast Two-Hybrid System,
Methods for Adeno-Associated Virus–Mediated Gene Transfer into Muscle,
Retroviral-Mediated Gene Transduction,
Gene Therapy Approaches to Sensitization of Human Prostate Carcinoma to Cisplatin by Adenoviral Expression of p53 and by Antisense
Oligonucleotide Methods,
Ribozyme Gene Therapy,
Index.