|
| G2892 Sigma |
Glass microcarrier beads90-150 μm, density: 1.04 g/cm3 at 25 °C |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |