![]() |
Properties
antibody form
purified immunoglobulin
clone
12H12
form
buffered aqueous solution
mol wt
antigen mol wt ~105 (with an additional band at 55 kDa)
species reactivity
human, hamster, bovine, chicken
does not react with
mouse, Xenopus
application(s)
immunoblotting: 0.5-1 μg/mL using HeLa cell extract
immunocytochemistry: suitable
immunoprecipitation: suitable
microarray: suitable
isotype
IgG1
Gene Information
human ... DDX20(11218)
shipped in
dry ice
storage temp.
−20°C
| ||||||||