Application
For clean up of glycan samples after reductive amination labeling or enzymatic digestion. The cartridge is equilibrated with acetonitrile, then loaded with a maximum of 20 μg of a glycan sample. The glycans are bound while excess dye and salts are removed by washing with acetonitrile. The glycans are then eluted with water.
Linkage
Recommended for use with the GlycoProfile 2-AA (Cat. No. PP0530) and 2-AB (Cat. No. PP0520) Labeling Kits.
Legal Information
GlycoProfile is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.