Components
A portable, self-contained unit for benchtop photography of electrophoresis gels.
Contains the following:
C0540 - Polaroid DS-34 Camera
Z359718 - Flat Face Hood, 7 inch (diagonal)
Z604747 - Gelcam Electrophoresis Filter Kit
General description
The Polaroid GelCam Camera system is used to make instant photographic records from light-emitting sources such as light boxes, video screens or CRT screens.
Features include:
• Hand-held operation
• No focusing required
• Four element f4.5 glass lens
• Shutter speeds: 1/125 sec to 1 sec and Bulb
• Easily interchangeable hoods
• Pistol grip single-stroke shutter release
• Rugged construction
• Snap-on hoods of various sizes determine the photographic area
Legal Information
GelCam is a registered trademark of Polaroid Corp.
Polaroid is a registered trademark of Polaroid Corp.