![]() |
Synonym
2
Molecular Formula
C2H7NO2S
Molecular Weight
109.15
CAS Number
MDL number
PubChem Substance ID
Descriptions
Biochem/physiol Actions
Antioxidant that binds hydroxyl radicals. Endogenous inhibitory neuromodulator that is blocked by strychnine and picrotoxin. Inhibits the Na2+-dependent transport of GABA and β-alanine. It is a degradation product of cysteine and a precursor of taurine.
Packaging
10 mg in autosmp vl
References
Reference
Green, T.R., et al., Antioxidant role and subcellular location of hypotaurine and taurine in human neutrophils. Biochim. Biophys. Acta 1073, 91-97, (1991) abstract
Ramanathan, V. K., Na+ -dependent gamma-aminobutyric acid (GABA) transport in the choroid plexus of rabbit. Biochim. Biophys. Acta 1330, 94, (1997) abstract
Brand, A., et al., Metabolism of cysteine in astroglial cells: synthesis of hypotaurine and taurine. J. Neurochem. 71, 827-832, (1998) abstract
Saransaari, P., and Oja, S.S., Uptake and release of β-alanine in cerebellar granule cells in primary culture: regulation of release by glutamatergic and GABAergic receptors. Neuroscience 53, 475-481, (1993) abstract
Okamoto, K., and Sakai, Y., Inhibitory actions of taurocyamine, hypotaurine, homotaurine, taurine and GABA on spike discharges of Purkinje cells. Brain Res. 206, 371-386, (1981) abstract
Tadolini, B., et al., Effects of taurine and hypotaurine on lipid peroxidation. Biochem. Biophys. Res. Commun. 213, 820-826, (1995) abstract
| ||||||||