![]() |
Properties
antibody form
affinity isolated antibody
form
buffered aqueous solution
species reactivity
human
application(s)
immunoblotting: 1:500 using whole extracts of human 293 cells or mouse NIH3T3 cells
microarray: suitable
Gene Information
human ... HAT1(8520)
shipped in
dry ice
storage temp.
−20°C
References
Reference
Sterner, D.E., and, Berger, S.L. Microbiol. Rev. 64, 435, (2000)
Roth, S.Y., et al. Annu. Rev. Biochem. 70, 81, (2001) abstract
| ||||||||