![]() |
Properties
antibody form
affinity isolated antibody
form
buffered aqueous solution
species reactivity
human, mouse, chicken
application(s)
immunoblotting: 1:2,500 using whole extracts of human 293 cells, mouse NIH3T3 cells, or chicken fibroblasts
Gene Information
human ... HAT1(8520)
mouse ... Hat1(107435)
shipped in
dry ice
storage temp.
−20°C
References
Reference
Sterner, D.E., and, Berger, S.L. Microbiol. Rev. 64, 435, (2000)
Roth, S.Y., et al. Annu. Rev. Biochem. 70, 81, (2001) abstract
| ||||||||