General description
This covers all basic aspects of human cytogenetic study other than malignancies and abnormalities. Since the 2nd edition, the largest new development has been the transition of fluorescence in situ hybridization to an essential tool for cytogenetecists. Its use in chromosome analysis is covered in detail. Another new technology described in detail is computerised image analysis. Conventional techniques have not been forgotten, with chapters on chromosome staining and banding techniques and meiotic studies. New authors take a fresh look at lymphocyte culture and prenatal diagnosis
Table of Contents
1. An introduction to human chromosomes and their analysis,
2. Lymphocyte culture, 3. The cytogenetics of pregnancy, 4. Chromosome staining and banding techniques, 5. The application of cytogenetic investigations to clinical practice, 6. The application of fluorescence in situ hybridization to chromosome analysis, 7. Meiotic studies in humans, 8. Microscopy and image analysis systems, Appendix 1: List of suppliers Appendix 2: Safety considerations Appendix 3: Chromosomes Index
| ||||||||